Learn More
Roboz Surgical SEMKEN MICRODISSECTING FORCEPS
SDP
Catalog No. NC9174752
Encompass
Semken Forceps; 1X2 Teeth; 1.2mm Tip Width; 5 in. Length
Catalog No. NC9174752 Supplier Roboz Surgical Supplier No. RS5242
Semken Forceps; 1X2 Teeth; 1.2mm Tip Width; 5 in. Length