Learn More
Med Vet International Miltex Vantage Semken Tissue Forceps, 5in
Catalog No. 502559461
Encompass_Preferred
The Miltex Vantage Semken Tissue Forceps are made of mid-grade stainless steel and have a length of 5in. They have a thumb handle and a straight tip with 1x2 teeth. These surgical forceps are commonly use to hold and manipulate delicate tissue.
Catalog No. 50-255-9461 Supplier Med Vet International Supplier No. MILV96106
The Miltex Vantage Semken Tissue Forceps are made of mid-grade stainless steel and have a length of 5in. They have a thumb handle and a straight tip with 1x2 teeth. These surgical forceps are commonly use to hold and manipulate delicate tissue.