Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Sigma Aldrich seMpai

Catalog No. 9022685MG Shop All MilliporeSigma Sigma Organic Chemistry Products
Encompass
Encompass_Preferred
Change view
Click to view available options
Quantity:
1 mg
5 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
90-226-85MG 5 mg
90-226-81MG 1 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 90-226-85MG Supplier Sigma Aldrich Supplier No. 9022685MG
Only null left
Add to Cart
Add to Cart

MilliporeSigma Sigma Organics products encompass a wide range of quality reagents, solvents, catalysts, and building blocks for organic synthesis. From benchtop discovery to process development and scale-up, Sigma Organics solutions are built to meet the needs of synthetic chemists.

2-[(1E,3E)-6-[3-(dimethylamino)pyridine]-1,3-butadien-1-yl]-4,5-dihydro-(4S)-4-thiazolecarboxylic acid sodium salt (DP3-Na) is a new NIR emission luciferin analog having luminescence peak at 670 ∼ 680 nm with a peak range in the near-infrared (NIR) region with the enhanced solubility in water.Luciferin is a generic term for the light-emitting compound found in organisms that generate bioluminescence.Applications:BioimagingIn vivo imaging

Specifications

Molecular Formula C15H16N3NaO2S
Linear Formula C15H16N3NaO2S
Quantity 5 mg
Synonym DP3-Na; TokeOni analogue; water-soluble NIR emission luciferin analogue; 2-[(1 E,3 E)-6-[3-(dimethylamino)pyridine]-1,3-butadien-1-yl]-4,5-dihydro-(4 S)-4-thiazolecarboxylic acid sodium salt
Molecular Weight (g/mol) 325.36
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.