Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Sempermed™ SemperForce™ Black Nitrile Gloves

Catalog No. 1904858A Shop All Sempermed USA Inc. Products
Change view
Click to view available options
Size:
Small
Medium
Large
X-Large
2X-Large
5 product options available for selection
Product selection table with 5 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Size
19-048-58A Small
19-048-58B Medium
19-048-58C Large
19-048-58D X-Large
19-048-58E 2X-Large
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 5 options available.
5 options
This item is not returnable. View return policy
Catalog No. 19-048-58A Supplier Sempermed™ Supplier No. BKNF102
Add to Cart
Add to Cart
This item is not returnable. View return policy

Black nitrile gloves feature textured fingertips to ensure secure gripping capabilities for demanding work environments.

  • Enhanced dexterity and gripping capabilities
  • Hides stains caused by grease, dirt, dyes and ink
  • ESD tested and approved
  • Ideal for industrial, electronics, automotive applications

Specifications

Certifications/Compliance FDA, ESD tested and approved
Applications Industrial
Color Black
Disposable Single-use
Exterior Finish Partially Textured
Latex Free Yes
Material Nitrile
Powdered Powder Free
Size Small
Thickness (English) 4.5 mil
Thickness (Metric) 0.11 mm
Model BKNF102
Length (English) 9 in.
Length (Metric) 23 cm
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.