Learn More
Ted Pella Inc Standard SEM Pin Stub Mount, Ø12.7mm x 8mm pin height, 100/pk
SDP
Catalog No. NC1678158
Encompass
FEI, Tescan, Zeiss (also for Philips, Aspex, Leo, Cambridge, AMRAY, Leica, CamScan, ETEC), aluminum, grooved edge
Catalog No. NC1678158 Supplier Ted Pella Inc Supplier No. 16111PK100
FEI, Tescan, Zeiss (also for Philips, Aspex, Leo, Cambridge, AMRAY, Leica, CamScan, ETEC), aluminum, grooved edge