Learn More
Abcam SEN 12333, alpha7 nicotinic acetylcholine receptor (nAChR) agonist, 10MG
Catalog No. 502706270
Encompass_Preferred
MW 339.4 Da, Purity >98%. Potent, selective α7 nicotinic acetylcholine receptor (nAChR) agonist. High affinity for rat α7 receptors (Ki = 260 nM). Full agonist in functional Ca²+ flux studies (EC₅₀ = 1.6 μM). Active in vivo and in vitro. Excellent brain penetration. Orally bioavailable.
The product is subject to the following: Abcam Restricted Use Statement
Catalog No. 50-270-6270 Supplier Abcam Supplier No. AB14223710MG
MW 339.4 Da, Purity >98%. Potent, selective α7 nicotinic acetylcholine receptor (nAChR) agonist. High affinity for rat α7 receptors (Ki = 260 nM). Full agonist in functional Ca²+ flux studies (EC₅₀ = 1.6 μM). Active in vivo and in vitro. Excellent brain penetration. Orally bioavailable.
The product is subject to the following: Abcam Restricted Use Statement