Learn More
Cayman Chemical Sennidin A, Purity: ≥98%, CAS: 641-12-3, 10 mg
SDP
Catalog No. 502909331
Encompass_Preferred
An active metabolite of sennoside A; formed from sennoside A by gut microbiota; increases fecal excretion in mice
Catalog No. 50-290-9331 Supplier Cayman Chemical Supplier No. 3523710 MG
An active metabolite of sennoside A; formed from sennoside A by gut microbiota; increases fecal excretion in mice