Learn More
Cayman Chemical SencIo cInrarIa MareIn 10mg
SDP
Catalog No. 502950815
Encompass_Preferred
A glucoside chalcone; scavenges DPPH radicals (IC50 = 48.35 μM); inhibits EGFR (IC50 = 19.94 μM); reduces fasting blood glucose and fasting serum insulin, triglycerides, LDL cholesterol, total cholesterol, IL-6, and CCL2 levels and renal IL-6 and CCL2 levels, as well as decreases fibrosis, in diabetic db/db mice at 50 mg/kg
Catalog No. 50-295-0815 Supplier Cayman Chemical Supplier No. 3321310 MG
A glucoside chalcone; scavenges DPPH radicals (IC50 = 48.35 μM); inhibits EGFR (IC50 = 19.94 μM); reduces fasting blood glucose and fasting serum insulin, triglycerides, LDL cholesterol, total cholesterol, IL-6, and CCL2 levels and renal IL-6 and CCL2 levels, as well as decreases fibrosis, in diabetic db/db mice at 50 mg/kg