Learn More
ATCC Sendai virus; Strain: Cantell
Catalog No. 502382406
Encompass
Sendai virus strain Cantell is propagated in SPF chicken embryonated eggs. This strain was deposited by the Central Public Health Laboratory, Helsinki, Finland. This product is whole-genome sequenced and is widely used for induction of interferon and cell fusion. It also has applications in infectious and respiratory disease research.
Catalog No. 50-238-2406 Supplier ATCC Supplier No. VR907
Sendai virus strain Cantell is propagated in SPF chicken embryonated eggs. This strain was deposited by the Central Public Health Laboratory, Helsinki, Finland. This product is whole-genome sequenced and is widely used for induction of interferon and cell fusion. It also has applications in infectious and respiratory disease research.