Learn More
ATCC Sendai virus; Strain: Sendai/52
Catalog No. 502384851
Encompass
Sendai virus strain Sendai/52 is propagated in SPF chicken embryonated eggs. This strain was isolated from the lung of a patient with a fatal case of newborn pneumonitis in Japan in 1952 and was deposited by the Centers for Disease Control and Prevention. The virus has applications in respiratory disease research.
Catalog No. 50-238-4851 Supplier ATCC Supplier No. VR105
Sendai virus strain Sendai/52 is propagated in SPF chicken embryonated eggs. This strain was isolated from the lung of a patient with a fatal case of newborn pneumonitis in Japan in 1952 and was deposited by the Centers for Disease Control and Prevention. The virus has applications in respiratory disease research.