Learn More
Enzo Life Sciences Sendide (1mg)
SDP
Supplier:  Enzo Life Sciences BMLP5100001
Encompass_Preferred
Potent NK1 neurokinin receptor antagonist. Alternative name: [Tyr6, D-Phe7, D-His9] Substance P (6-11). Purity: ≥97%. Solubility: Soluble in Water. Long Term Storage: -20°C.
Catalog No. 50-201-1360