Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC Senecionine N-oxide | 13268-67-2 | 1 MG
SDP

Catalog No. 5000419466
Encompass_Preferred

Senecionine N-oxide is the primary product of pyrrolizidine alkaloid biosynthesis found in root cultures of *Senecio vulgaris*. This compound exhibits anti-cancer activity.

  • Molecular weight: 351.39
  • Molecular formula: C18H25NO6
  • Appearance: Solid
  • Color: White to off-white
  • Purity: 99.11%
  • Solubility: 50 mg/mL in H2O (requires ultrasonic)
  • Biological activity: Cytotoxicity against human A204 cells (IC50: 150 μg/mL)
  • Shipping: Room temperature in continental US
  • Storage: 4°C, protected from light (for solid); -80°C for 6 months; -20°C for 1 month in solvent (protected from light)

Catalog No. 50-004-19466 Supplier Medchemexpress LLC Supplier No. HYN66371MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Senecionine N-oxide is the primary product of pyrrolizidine alkaloid biosynthesis found in root cultures of *Senecio vulgaris*. This compound exhibits anti-cancer activity.

  • Molecular weight: 351.39
  • Molecular formula: C18H25NO6
  • Appearance: Solid
  • Color: White to off-white
  • Purity: 99.11%
  • Solubility: 50 mg/mL in H2O (requires ultrasonic)
  • Biological activity: Cytotoxicity against human A204 cells (IC50: 150 μg/mL)
  • Shipping: Room temperature in continental US
  • Storage: 4°C, protected from light (for solid); -80°C for 6 months; -20°C for 1 month in solvent (protected from light)

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.