Learn More
Medchemexpress LLC Senecionine N-oxide | 13268-67-2 | 5 MG
SDP
Catalog No. 5000417988
Encompass_Preferred
Senecionine N-oxide is the primary product of pyrrolizidine alkaloid biosynthesis in root cultures of *Senecio vulgaris*. It has anti-cancer activity and is for research use only.
  • Exhibits anti-cancer activity
  • Primary product of pyrrolizidine alkaloid biosynthesis
  • Suitable for research applications
Catalog No. 50-004-17988 Supplier Medchemexpress LLC Supplier No. HYN66375MG
Senecionine N-oxide is the primary product of pyrrolizidine alkaloid biosynthesis in root cultures of *Senecio vulgaris*. It has anti-cancer activity and is for research use only.
  • Exhibits anti-cancer activity
  • Primary product of pyrrolizidine alkaloid biosynthesis
  • Suitable for research applications