Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC Senecionine (senecionan-11,16-dione, 12-hydroxy-) | 130-01-8 | MFCD00221720 | 99.5% | 335.39 g/mol | C18H25NO5 | 10 MG
SDP

Catalog No. 5000289641
Encompass_Preferred

Senecionine is a pyrrolizidine alkaloid supplied as an analytical standard for research and analytical applications. The compound is provided as a solid and has been reported to decrease glutathione S-transferase activity and related enzymatic activities. Handle and store protected from light.

  • Analytical standard for research and analytical applications.
  • High purity: 99.5%.
  • CAS number 130-01-8.
  • Molecular formula C18H25NO5 and molecular weight 335.39 g/mol.
  • Supplied as a 10 MG solid, white to off-white.
  • Store protected from light; in solvent: -80°C (6 months), -20°C (1 month).

Catalog No. 50-002-89641 Supplier Medchemexpress LLC Supplier No. HYN256010MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Senecionine is a pyrrolizidine alkaloid supplied as an analytical standard for research and analytical applications. The compound is provided as a solid and has been reported to decrease glutathione S-transferase activity and related enzymatic activities. Handle and store protected from light.

  • Analytical standard for research and analytical applications.
  • High purity: 99.5%.
  • CAS number 130-01-8.
  • Molecular formula C18H25NO5 and molecular weight 335.39 g/mol.
  • Supplied as a 10 MG solid, white to off-white.
  • Store protected from light; in solvent: -80°C (6 months), -20°C (1 month).

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.