Learn More
Medchemexpress LLC HY-50694 100mg , Senicapoc ICA-17043 CAS:289656-45-7 Purity:98%
SDP
Supplier:  Medchemexpress LLC HY50694100MG
Encompass_Preferred
Medchemexpress, HY-50694 100mg Senicapoc ICA-17043 CAS:289656-45-7 Formula:C20H15F2NO IC50: 11 nM (Gardos channel) Purity:98% Senicapoc is a potent and selective Gardos channel blocker with IC 50 value of 11 nM, and blocks Ca 2+ -induced rubidium flux from human RBCs with an IC 50 value of 11 nM and inhibits RBC dehydration with IC 50 of 30 nM. Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-149-2715