Learn More
Eurofins Discoverx Senktide, [Succinyl-Asp6, NMephe8]-Substance P (6-11), 1 mg. (92-1066)
Supplier:  Eurofins Discoverx 921066
Encompass
Senktide, [Succinyl-Asp6, nMephe8]-Substance P (6-11) is a neuropeptide and substance P analog. It is a selective neurokinin NK3 tachykinin receptor agonist, which causes dopamine neuron excitation, enhanced dopaminergic function, increased locomotion, and may be linked to psychological disorders. Provided as 1 mg of dry powder alongside reconstitution buffer. These highly purified, pre-validated control ligands have been used to qualify our InCELL Hunter,  PathHunter and cAMP Hunter cell lines as well as our Assay Ready Kits. Ligands are conveniently supplied with a recommended reconstitution buffer. For detailed information and product-specific datasheet, visit www.discoverx.com/ligands.
Catalog No. 50-199-9607