Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC Senktide | 106128-89-6 | 99.9% | 841.97 | 50 MG
SDP

Catalog No. 5000426267
Encompass_Preferred

Senktide is a potent and selective agonist of the neuromedin K3 (NK3) receptor, with an EC50 ranging from 0.5-3 nM. It also agonizes the NK1 receptor, though less potently (EC50=35 μM). This compound shows promise in research related to neurological diseases.

  • Highly potent and selective: demonstrates strong agonistic activity towards the NK3 receptor, with significantly lower potency for the NK1 receptor.
  • Neurological research focus: promising for use in the study of neurological diseases.
  • Comprehensive data available: detailed information on cellular effects, in vitro, and in vivo studies are provided.
  • Multiple package sizes: available in various quantities from 1 mg to 500 mg to suit different research needs.
  • High purity: the product boasts a purity of 99.91%.

Catalog No. 50-004-26267 Supplier Medchemexpress LLC Supplier No. HYP018750MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Senktide is a potent and selective agonist of the neuromedin K3 (NK3) receptor, with an EC50 ranging from 0.5-3 nM. It also agonizes the NK1 receptor, though less potently (EC50=35 μM). This compound shows promise in research related to neurological diseases.

  • Highly potent and selective: demonstrates strong agonistic activity towards the NK3 receptor, with significantly lower potency for the NK1 receptor.
  • Neurological research focus: promising for use in the study of neurological diseases.
  • Comprehensive data available: detailed information on cellular effects, in vitro, and in vivo studies are provided.
  • Multiple package sizes: available in various quantities from 1 mg to 500 mg to suit different research needs.
  • High purity: the product boasts a purity of 99.91%.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.