Learn More
Enzo Life Sciences Senktide (1mg)
SDP
Supplier:  Enzo Life Sciences BMLP5090001
Encompass_Preferred
Highly selective NK3 neurokinin receptor agonist. Alternative name: [Succinyl-Asp6,N-Me-Phe8] Substance P (6-11). Purity: ≥97%. Solubility: Methanol(25 mg/ml), Ethanol (25 mg/ml), DMSO (25 mg/ml). Long Term Storage: -20°C.
Catalog No. 50-201-1359