Learn More
Medchemexpress LLC Senkyunolide A | 63038-10-8 | 192.26 | 5 MG
SDP
Catalog No. 5000416830
Encompass_Preferred
Senkyunolide A is a phthalide with anticancer activity, acting as an anti-tumor cell proliferation agent. It offers neuroprotective effects by decreasing protein phosphatase PP2A and α-synuclein phosphorylation, and inhibits osteoarthritis by modulating the NLRP3 signaling pathway. Furthermore, it suppresses the expression of CD137, a biomarker for atherosclerosis.
  • Inhibits proliferation of HT-29 and CCD-18Co cells.
  • Decreases high expression of CD137 in TNF-α treated MAECs cells.
  • Enhances cell viability and reduces apoptosis in PC12 cells.
  • Stimulates cell proliferation and curbs apoptosis in IL-1β-treated chondrocytes.
  • Inhibits cell inflammation by blocking the NLRP3 signaling pathway.
  • Demonstrates improvement in mouse models of osteoarthritis, alleviating articular cartilage destruction.
Catalog No. 50-004-16830 Supplier Medchemexpress LLC Supplier No. HYN07435MG
Senkyunolide A is a phthalide with anticancer activity, acting as an anti-tumor cell proliferation agent. It offers neuroprotective effects by decreasing protein phosphatase PP2A and α-synuclein phosphorylation, and inhibits osteoarthritis by modulating the NLRP3 signaling pathway. Furthermore, it suppresses the expression of CD137, a biomarker for atherosclerosis.
  • Inhibits proliferation of HT-29 and CCD-18Co cells.
  • Decreases high expression of CD137 in TNF-α treated MAECs cells.
  • Enhances cell viability and reduces apoptosis in PC12 cells.
  • Stimulates cell proliferation and curbs apoptosis in IL-1β-treated chondrocytes.
  • Inhibits cell inflammation by blocking the NLRP3 signaling pathway.
  • Demonstrates improvement in mouse models of osteoarthritis, alleviating articular cartilage destruction.