Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC Senkyunolide A | 63038-10-8 | MFCD16660665 | 99.3% | 192.26 g/mol | C12H16O2 | 10 MG
SDP

Catalog No. 5000296137
Encompass_Preferred

Senkyunolide A is a phthalide-class natural product supplied as a research-grade analytical standard, reported to have anticancer and neuroprotective activities and commonly used in biochemical and cellular studies.

  • High purity (~99.3%), suitable for analytical and biological research.
  • Molecular formula C12H16O2, molecular weight 192.26 g/mol.
  • Available in small research quantities (e.g., 10 MG) for lab-scale experiments.
  • Appearance: colorless to light yellow liquid; density ~1.1 g/cm3.
  • Storage: store at -80°C, protect from light, under inert gas when required.

Catalog No. 50-002-96137 Supplier Medchemexpress LLC Supplier No. HYN074310MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Senkyunolide A is a phthalide-class natural product supplied as a research-grade analytical standard, reported to have anticancer and neuroprotective activities and commonly used in biochemical and cellular studies.

  • High purity (~99.3%), suitable for analytical and biological research.
  • Molecular formula C12H16O2, molecular weight 192.26 g/mol.
  • Available in small research quantities (e.g., 10 MG) for lab-scale experiments.
  • Appearance: colorless to light yellow liquid; density ~1.1 g/cm3.
  • Storage: store at -80°C, protect from light, under inert gas when required.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.