Learn More
Selleck Chemical LLC Senkyunolide I S3275-1mg
SDP
Catalog No. 503109110
Encompass_Preferred
Senkyunolide I (SEI SENI) is an orally active compound isolated from Ligusticum chuanxiong with analgesic anti-migraine neuroprotective anti-oxidation and anti-apoptosis activities Senkyunolide I (SEI SENI) up-regulates the phosphorylation of Erk1/2 and induces Nrf2 nuclear translocation with enhanced HO-1 and NQO1 expressions Senkyunolide I (SEI SENI) promotes the ratio of Bcl-2/Bax and inhibits the expressions of cleaved caspase 3 and caspase 9
Catalog No. 50-310-9110 Supplier Selleck Chemical LLC Supplier No. S32751MG
Add to Cart
Add to Cart
Senkyunolide I (SEI SENI) is an orally active compound isolated from Ligusticum chuanxiong with analgesic anti-migraine neuroprotective anti-oxidation and anti-apoptosis activities Senkyunolide I (SEI SENI) up-regulates the phosphorylation of Erk1/2 and induces Nrf2 nuclear translocation with enhanced HO-1 and NQO1 expressions Senkyunolide I (SEI SENI) promotes the ratio of Bcl-2/Bax and inhibits the expressions of cleaved caspase 3 and caspase 9