Learn More
Medchemexpress LLC HY-N6935 5mg Medchemexpress, Sennidin B CAS:517-44-2 Purity:>98%
SDP
Supplier:  Medchemexpress LLC HYN69355MG
Encompass_Preferred
Medchemexpress, HY-N6935 5mg Sennidin B CAS:517-44-2 Sennidin B, a stereoisomer isolated from the leaves of Cassia angustifolia, has lower activity than Sennidin A. Sennidin A inhibits HCV NS3 helicase, with an IC50 of 0.8 μM. Sennidin A induces phosphorylation of Akt and glucose transporter 4 (GLUT4) translocation. Sennidin A stimulates the glucose incorporation . Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-208-2973