Learn More
Medchemexpress LLC HY-N0365 5mg Medchemexpress, Sennoside A CAS:81-27-6 Purity:>98%
SDP
Supplier:  Medchemexpress LLC HYN03655MG
Encompass_Preferred
Medchemexpress, HY-N0365 5mg Sennoside A CAS:81-27-6 Sennoside A is an anthraquinone glycoside, found in large quantities in leaves and pods of Senna (Cassia angustifolia). Sennoside A is a HIV-1 inhibitor effective on HIV-1 replication. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-197-0258