Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Sennoside C, ≥98% (HPLC), MilliporeSigma™ Supelco™

Catalog No. 111014075 Shop All MilliporeSigma Supelco Products
Change view
Click to view available options
Quantity:
10 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
11-101-4075 10 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 11-101-4075 Supplier MilliporeSigma Supelco Supplier No. 7323510MG
Add to Cart
Add to Cart

Sennoside C is an anthraquinone glycoside, belonging to the group of sennosides, widely found in Senna plant. It is commonly used worldwide as a natural laxative.

Food and Beverages
TRUSTED_SUSTAINABILITY

Specifications

Packaging Bottomless Glass Bottle with Inserted Fused Cone
CAS 37271-16-2
Flash Point 344.4°C
Physical Form Neat
Linear Formula C42H40O19
Type Analytical Standard
Quantity 10 mg
Formula Weight 848.76
Percent Purity ≥98% (HPLC)
Grade Analytical Standard
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.