Learn More
TARGETMOL CHEMICALS INC Sennoside D 5MG
SDP
Catalog No. 502879274
Encompass_Preferred
Also available in 1 mL, 1 mg, 10 mg, 25 mg, 50 mg and bulk. Please contact Fisher for quotes. Sennoside D is an anthraquinone glycoside extracted from leaves and pods of Cassia angustifolia. Purity 99.44%
Catalog No. 50-287-9274 Supplier TARGETMOL CHEMICALS INC Supplier No. T2S09685MG
Also available in 1 mL, 1 mg, 10 mg, 25 mg, 50 mg and bulk. Please contact Fisher for quotes. Sennoside D is an anthraquinone glycoside extracted from leaves and pods of Cassia angustifolia. Purity 99.44%