Learn More
Cayman Chemical Sennoside B, Purity: ≥98%, CAS: 128-57-4, 50 mg
SDP
Catalog No. 502938762
Encompass_Preferred
A glycoside; inhibits H+/K+-ATPase activity in isolated rat stomach mucosa and increases levels of PGE2 in AGS gastric cells at 100 µM; increases the intestinal transport rate by 72.2% but has no effect on the gastric emptying rate in mice at 100 mg/kg; reduces lesion indices by 39.9 and 62.9% in HCl/ethanol-induced gastritis and indomethacin-induced gastric ulcers, respectively, in rats at 100 mg/kg
Catalog No. 50-293-8762 Supplier Cayman Chemical Supplier No. 2841050 MG
A glycoside; inhibits H+/K+-ATPase activity in isolated rat stomach mucosa and increases levels of PGE2 in AGS gastric cells at 100 µM; increases the intestinal transport rate by 72.2% but has no effect on the gastric emptying rate in mice at 100 mg/kg; reduces lesion indices by 39.9 and 62.9% in HCl/ethanol-induced gastritis and indomethacin-induced gastric ulcers, respectively, in rats at 100 mg/kg