Learn More
Enzo Life Sciences SENP2 (catalytic domain) (human), (recombinant) (GST-tag) (100 µg)
SDP
Supplier:  Enzo Life Sciences BMLUW97650100
Encompass_Preferred
Alternative name: Sentrin specific protease 2. Purity: ≥90% (densitometry). Formulation: Liquid. In 50mM TRIS, pH 7.5, containing 150mM sodium chloride, 10% glycerol, and 0.5mM DTT. Source: Produced in E. coli. Specificity: Demonstrated recognition of SUMO-1 and SUMO-2 in vitro. Long Term Storage: -80°C. Applications: Activity assay.
Catalog No. 50-201-2330