Learn More
Aviva Systems Biology Corp Senp6 Antibody - C-terminal region (ARP50599_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP50599_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against Senp6. It was validated on Western Blot. . Alternate names: 2810017C20Rik, E130319N12Rik, MGC38009, Susp1, mKIAA0797ApplicationWB. Species reactivity: Cow, Dog, Horse, Human, Mouse, Pig, Rabbit, Rat
Catalog No. 50-285-518