Learn More
BioVendor, LLC QuickZyme Sensitive Tissue Collagen Assay Kit 5 X 96-Assays
SDP
Catalog No. 502069128
Encompass
QuickZyme Sensitive Tissue Collagen Assay Kit, 5 X 96-Assays. Measure hydroxyproline in small tissue samples with low collagen levels! The ONLY hydroxyproline-based assay which shows no matrix effect and no need for further dilution of the hydrolysate, enabling measurement in small samples with low hydroxyproline levels, such as liver. No drying of hydrolysate is needed, resulting in a fast (<2 hrs), sensitive and easy (96-well plate format) assay. Dynamic range: 6-300 ug/ml. Sensitivity: 2.5 ug/ml. Sample vol. 50-500 ul. Species independent. Measures total hydroxyproline. If this hydroxyproline is obtained upon hydrolysis of collagen, it represents all collagen types present without discriminating between types of collagen, procollagen, mature collagen and collagen degradation products. Hydroxyproline in tissue hydrolysates can be used as a direct measure of the amount of collagen present. Collagen hydrolysis tubes & protocol ARE included. Ease-of-use: Equivalent to ELISA.
Catalog No. 50-206-9128 Supplier BioVendor, LLC Supplier No. QZBTISCOL5
QuickZyme Sensitive Tissue Collagen Assay Kit, 5 X 96-Assays. Measure hydroxyproline in small tissue samples with low collagen levels! The ONLY hydroxyproline-based assay which shows no matrix effect and no need for further dilution of the hydrolysate, enabling measurement in small samples with low hydroxyproline levels, such as liver. No drying of hydrolysate is needed, resulting in a fast (<2 hrs), sensitive and easy (96-well plate format) assay. Dynamic range: 6-300 ug/ml. Sensitivity: 2.5 ug/ml. Sample vol. 50-500 ul. Species independent. Measures total hydroxyproline. If this hydroxyproline is obtained upon hydrolysis of collagen, it represents all collagen types present without discriminating between types of collagen, procollagen, mature collagen and collagen degradation products. Hydroxyproline in tissue hydrolysates can be used as a direct measure of the amount of collagen present. Collagen hydrolysis tubes & protocol ARE included. Ease-of-use: Equivalent to ELISA.