Learn More
ILEX LIFE SCIENCES LLC QuickZyme Sensitive Tissue Hydroxyproline Kit, 5 x 96 Assays Bulk Pack, 96-well plate format, colorimetric assay for low level collagen analysis (tissue, tissue homogenates samples)
SDP
Catalog No. 502120039
Encompass_Preferred
QuickZyme Sensitive Tissue Hydroxyproline Assay Kit enables collagen analysis in small tissue samples with low collagen levels. Quantitative colorimetric determination of hydroxyproline residues, obtained by acid hydrolysis of collagen. The only hydroxyproline-based assay which shows no matrix effect and no need for further dilution of hydrolysate. No drying of hydrolysate is needed, resulting in a fast and easy (96-well plate format) assay. NOTE: Kit does not contain tubes and protocol for collagen hydrolysis. If you need these items please order the QuickZyme Sensitive Tissue Collagen Assay. Application: tissues with low collagen/hydroxyproline content (ex. liver). Sensitivity: 2.4 µM (0.3 µg/ml). Assay-time: < 2 hours. Collagen is a main component of extracellular matrix. Dysregulation in collagen production results in pathologies such as fibrosis (too much collagen), or osteoarthritis (too little collagen). Thus measurement of collagen production is important in many studies.
Catalog No. 50-212-0039 Supplier ILEX LIFE SCIENCES LLC Supplier No. QZBTISHYP5
QuickZyme Sensitive Tissue Hydroxyproline Assay Kit enables collagen analysis in small tissue samples with low collagen levels. Quantitative colorimetric determination of hydroxyproline residues, obtained by acid hydrolysis of collagen. The only hydroxyproline-based assay which shows no matrix effect and no need for further dilution of hydrolysate. No drying of hydrolysate is needed, resulting in a fast and easy (96-well plate format) assay. NOTE: Kit does not contain tubes and protocol for collagen hydrolysis. If you need these items please order the QuickZyme Sensitive Tissue Collagen Assay. Application: tissues with low collagen/hydroxyproline content (ex. liver). Sensitivity: 2.4 µM (0.3 µg/ml). Assay-time: < 2 hours. Collagen is a main component of extracellular matrix. Dysregulation in collagen production results in pathologies such as fibrosis (too much collagen), or osteoarthritis (too little collagen). Thus measurement of collagen production is important in many studies.