Learn More
Sensidyne™ Gilian™ LFS-113 Air Sampling System
SDP
Catalog No. 19087531 Shop All Sensidyne Inc. Products
Encompass_Preferred
Change view
Click to view available options
With Clock
Without Clock
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No.
19-087531 Without Clock
19-087532 With Clock
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 19-087531 Supplier Sensidyne™ Supplier No. 800088111
Item Discontinued This item has been discontinued by the supplier. Please to view product availability in your area.
Includes
Rechargeable NiCd battery pack (included) provides eight hours of continuous operation.
Dual-mode, low-flow air sampler
Personal low-flow air samplers ideal for sorbent tube collection of gases and vapors (18-556V and 18-556W sorbent tubes sold separately). Feature two modes of operation: constant- flow mode (5 to 200cc/min.) for single-tube sampling and constant-pressure mode (1 to 350cc/min.) for multiple-tube sampling.
Instant fault function automatically stops internal pump when sampler is operated outside its performance envelope. EMI/RFI shielding case protects instrument from electromagnetic and radio-frequency interference.
Order Info
Compact, lightweight unit features belt clip. Sampler measures approximately 2 1/2L x 1 1/2W x 4 1/2 in.H (6.4 x 3.6 x 11.7cm). Net weight: 12 oz. (340g). Unit is available with or without a time-elapsed clock.
Certifications
UL listed, UL listed as intrinsically safe for use in Class I, Groups A, B, C, and D; Class II, Groups E, F, and G; and Class III hazardous locations.