Learn More
Sensidyne™ Gilian™ LFS-113 Air Sampling System
SDP
Catalog No. 19087532 Shop All Sensidyne Inc. Products
Encompass_Preferred
Change view
Click to view available options
With Clock
Without Clock
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No.
19-087532 With Clock
19-087531 Without Clock
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 19-087532 Supplier Sensidyne™ Supplier No. 800098111
Includes
Rechargeable NiCd battery pack (included) provides eight hours of continuous operation.
Dual-mode, low-flow air sampler
Personal low-flow air samplers ideal for sorbent tube collection of gases and vapors (18-556V and 18-556W sorbent tubes sold separately). Feature two modes of operation: constant- flow mode (5 to 200cc/min.) for single-tube sampling and constant-pressure mode (1 to 350cc/min.) for multiple-tube sampling.
Instant fault function automatically stops internal pump when sampler is operated outside its performance envelope. EMI/RFI shielding case protects instrument from electromagnetic and radio-frequency interference.
Order Info
Compact, lightweight unit features belt clip. Sampler measures approximately 2 1/2L x 1 1/2W x 4 1/2 in.H (6.4 x 3.6 x 11.7cm). Net weight: 12 oz. (340g). Unit is available with or without a time-elapsed clock.
Certifications
UL listed, UL listed as intrinsically safe for use in Class I, Groups A, B, C, and D; Class II, Groups E, F, and G; and Class III hazardous locations.