Learn More
Meridian Life Science SensiFAST Probe Lo-ROX Kit, 5000 x 20 μL reactions, TaqMan/Scorpions/molecular beacon compatible, Lo-ROX dye, antibody-mediated hot-start DNA polymerase, 2x mastermix
SDP
Catalog No. NC1665860
Encompass_Preferred
SensiFAST Probe Lo-ROX Kit, 5000 x 20 μL reactions, TaqMan/Scorpions/molecular beacon compatible, Lo-ROX dye, antibody-mediated hot-start DNA polymerase, 2x mastermix
Catalog No. NC1665860 Supplier Meridian Life Science Supplier No. BIO84050
SensiFAST Probe Lo-ROX Kit, 5000 x 20 μL reactions, TaqMan/Scorpions/molecular beacon compatible, Lo-ROX dye, antibody-mediated hot-start DNA polymerase, 2x mastermix