Learn More
Meridian Life Science SensiFAST SYBR No-ROX Mix 2X, 20 mL total, 2000 reactions, -20 °C storage, DNA RNA template compatible, antibody-mediated hot-start
SDP
Catalog No. NC0445206
Encompass
SensiFAST SYBR No-ROX Mix 2X, 20 mL total, 2000 reactions, -20 °C storage, DNA RNA template compatible, antibody-mediated hot-start
Catalog No. NC0445206 Supplier Meridian Life Science Supplier No. BIO98020
SensiFAST SYBR No-ROX Mix 2X, 20 mL total, 2000 reactions, -20 °C storage, DNA RNA template compatible, antibody-mediated hot-start