Learn More
Hach Company Sension 5071 Universal Cond
Catalog No. 5000207872 Shop All Hach Company Products
Encompass
Sension Plus 5071 Universal Conductivity cell for very low conductivity measurements in lab Conductivity cell with Pt electrodes and glass body for Sension Plus benchtop meters Cell constant k is 01 cm-1 Built-in Pt1000 for automatic temperature compensation 1m cable Measuring range 0 05-30 micro siemens/cm -30 - 85C The electrode is suitable for low conductivity samples purified and demineralised waters etc
Catalog No. 50-002-07872 Supplier Hach Company Supplier No. LZW5071.97.0002
Sension Plus 5071 Universal Conductivity cell for very low conductivity measurements in lab Conductivity cell with Pt electrodes and glass body for Sension Plus benchtop meters Cell constant k is 01 cm-1 Built-in Pt1000 for automatic temperature compensation 1m cable Measuring range 0 05-30 micro siemens/cm -30 - 85C The electrode is suitable for low conductivity samples purified and demineralised waters etc