Learn More
Hach Company Sension 5208 pH liquid filled
Catalog No. 5000207627 Shop All Hach Company Products
Encompass
Sension Plus 5208 pH liquid filled combination electrode for Micro Sample volumes-3 mm diameter-screw cap S7 pH liquid electrode (refillable) with ceramic diaphragm Glass body Range 0-14 pH at 0 to 80C Screw cap S7 3 mm diameter 5 mm minimum immersion depth This pH electrode is suitable for Micro Sample volumes
Catalog No. 50-002-07627 Supplier Hach Company Supplier No. LZW5208.97.0002
Sension Plus 5208 pH liquid filled combination electrode for Micro Sample volumes-3 mm diameter-screw cap S7 pH liquid electrode (refillable) with ceramic diaphragm Glass body Range 0-14 pH at 0 to 80C Screw cap S7 3 mm diameter 5 mm minimum immersion depth This pH electrode is suitable for Micro Sample volumes