Learn More
Hach Company Sension 5209 pH liquid filled
Catalog No. 5000207321 Shop All Hach Company Products
Encompass
Sension Plus 5209 pH liquid filled combination electrode for Micro Sample volumes-6 mm diameter-screw cap S7 pH liquid electrode (refillable) with ceramic diaphragm Glass body Range 0-14 pH at 0 to 80C Screw cap S7 6 mm diameter 8 mm minimum immersion depth This pH electrode is suitable for Micro Sample volumes
Catalog No. 50-002-07321 Supplier Hach Company Supplier No. LZW5209.97.0002
Sension Plus 5209 pH liquid filled combination electrode for Micro Sample volumes-6 mm diameter-screw cap S7 pH liquid electrode (refillable) with ceramic diaphragm Glass body Range 0-14 pH at 0 to 80C Screw cap S7 6 mm diameter 8 mm minimum immersion depth This pH electrode is suitable for Micro Sample volumes