Learn More
Hach Company Sension 5292 Cond cell
Catalog No. 5000207642 Shop All Hach Company Products
Encompass
Sension Plus 5292 Conductivity cell with two Platinum plates for general measurements in lab Conductivity cell with two Platinum plate electrodes and glass body for Sension Plus Laboratory Meters Cell constant k is 1 cm-1 1 m cable Measuring range 100 micro siemens/cm at 10-80C
Catalog No. 50-002-07642 Supplier Hach Company Supplier No. LZW5292.97.0002
Sension Plus 5292 Conductivity cell with two Platinum plates for general measurements in lab Conductivity cell with two Platinum plate electrodes and glass body for Sension Plus Laboratory Meters Cell constant k is 1 cm-1 1 m cable Measuring range 100 micro siemens/cm at 10-80C