Learn More
Hach Company Sension 5293 Platinum Conduct
Catalog No. 5000207622 Shop All Hach Company Products
Encompass
Sension Plus 5293 Platinum Conductivity cell for general measurements in lab Conductivity cell with 3 Platinum electrodes and glass body for Sension Plus Laboratory Meters Cell constant k is 0 7 cm-1 Built-in Pt1000 for automatic temperature compensation 1 m cable Measuring range 0 2 to 100 micro siemens/cm at 30-85C
Catalog No. 50-002-07622 Supplier Hach Company Supplier No. LZW5293.97.0002
Sension Plus 5293 Platinum Conductivity cell for general measurements in lab Conductivity cell with 3 Platinum electrodes and glass body for Sension Plus Laboratory Meters Cell constant k is 0 7 cm-1 Built-in Pt1000 for automatic temperature compensation 1 m cable Measuring range 0 2 to 100 micro siemens/cm at 30-85C