Learn More
Hach Company Sension pH comb electr 5015T
Catalog No. 5000207640 Shop All Hach Company Products
Encompass
Sension Plus pH liquid combination electrode 5015T for chemical pharma applications pH combination electrode (refillable) with 3 ceramic diaphragms encapsulated reference with silver ion barrier and built-in fast responding Pt1000 for Automatic Temperature Compensation KCl 3M glycerine electrolyte Glass body Range 0-14 pH -10 to 100C 1 m cable For Laboratory Meter This pH electrode is perfect for sample investigation samples containing proteins and for samples containing sulphurs reducing sugars or other substances which react with silver ions Very fast stabilization in rapid changes of sample temperature due to unique ConATC design
Catalog No. 50-002-07640 Supplier Hach Company Supplier No. LZW5015T.97.002
Sension Plus pH liquid combination electrode 5015T for chemical pharma applications pH combination electrode (refillable) with 3 ceramic diaphragms encapsulated reference with silver ion barrier and built-in fast responding Pt1000 for Automatic Temperature Compensation KCl 3M glycerine electrolyte Glass body Range 0-14 pH -10 to 100C 1 m cable For Laboratory Meter This pH electrode is perfect for sample investigation samples containing proteins and for samples containing sulphurs reducing sugars or other substances which react with silver ions Very fast stabilization in rapid changes of sample temperature due to unique ConATC design