Learn More
Hach Company Sension pH comb punct 5053T
Catalog No. 5000207883 Shop All Hach Company Products
Encompass
Sension Plus pH combination puncture electrode 5053T for food applications-automatic temperature compensation-for portable meters pH puncture electrode with built-in Automatic temperature compensation Pt 1000 for portable meters solid gel filled encapsulated Ag/AgCl reference 1 ceramic and 1 open diaphragm Glass electrode 316 stainless steel body Range 2-14 pH 0-60C 1m cable MP5 connector The electrode is suitable for food applications e g cheese meat fish fruits bread doughs etc
Catalog No. 50-002-07883 Supplier Hach Company Supplier No. LZW5053T.97.002
Sension Plus pH combination puncture electrode 5053T for food applications-automatic temperature compensation-for portable meters pH puncture electrode with built-in Automatic temperature compensation Pt 1000 for portable meters solid gel filled encapsulated Ag/AgCl reference 1 ceramic and 1 open diaphragm Glass electrode 316 stainless steel body Range 2-14 pH 0-60C 1m cable MP5 connector The electrode is suitable for food applications e g cheese meat fish fruits bread doughs etc