Learn More
Hach Company Sension pH comb punct 5233
Catalog No. 5000207866 Shop All Hach Company Products
Encompass
Sension Plus pH combination puncture electrode 5233 for Food ApplicationspH combination puncture electrode 5233 with one ceramic and one open diaphragm and solid electrolyte POM body Range 0 14 pH at 0-80C S7 screw cap This electrode is suitable for semisolid food samples e g cheeses meats fish fruits bread doughs etc The encapsulated reference ensures high stability and long lifetime
Catalog No. 50-002-07866 Supplier Hach Company Supplier No. LZW5233.97.0002
Sension Plus pH combination puncture electrode 5233 for Food ApplicationspH combination puncture electrode 5233 with one ceramic and one open diaphragm and solid electrolyte POM body Range 0 14 pH at 0-80C S7 screw cap This electrode is suitable for semisolid food samples e g cheeses meats fish fruits bread doughs etc The encapsulated reference ensures high stability and long lifetime