Learn More
Hach Company Sension pH combination electr
Catalog No. 5000207322 Shop All Hach Company Products
Encompass
Sension Plus pH combination electrode with screw cap S7 (for samples containing proteins) pH combination electrode with 3 ceramic diaphragms and refill (KCl glycerine) electrolyte Glass body Range 0-14 pH at 0-100C Screw cap S7 The electrode is suitable for samples containing proteins and also for high temperatures The electrode has a large semi-spheric membrane is very robust and fast responding
Catalog No. 50-002-07322 Supplier Hach Company Supplier No. LZW5211.97.0002
Sension Plus pH combination electrode with screw cap S7 (for samples containing proteins) pH combination electrode with 3 ceramic diaphragms and refill (KCl glycerine) electrolyte Glass body Range 0-14 pH at 0-100C Screw cap S7 The electrode is suitable for samples containing proteins and also for high temperatures The electrode has a large semi-spheric membrane is very robust and fast responding