Learn More
Hach Company Sension pH combination electr
Catalog No. 5000207886 Shop All Hach Company Products
Encompass
Sension Plus pH combination electrode 5221 with sleeve diaphragm and screw cap S7 for creams-viscous and dirty samples pH combination electrode 5221 with sleeve diaphragm and refillable electrolyte Glass body range 0-12 pH at 0-60C screw cap S7 This electrode is suitable for the most difficult samples e g for low ionic strength samples creams emulsions very viscous samples wines paint and samples with colloids The encapsulated reference ensures high stability and long lifetime The large cylindrical pH membrane provides fast response
Catalog No. 50-002-07886 Supplier Hach Company Supplier No. LZW5221.97.0002
Sension Plus pH combination electrode 5221 with sleeve diaphragm and screw cap S7 for creams-viscous and dirty samples pH combination electrode 5221 with sleeve diaphragm and refillable electrolyte Glass body range 0-12 pH at 0-60C screw cap S7 This electrode is suitable for the most difficult samples e g for low ionic strength samples creams emulsions very viscous samples wines paint and samples with colloids The encapsulated reference ensures high stability and long lifetime The large cylindrical pH membrane provides fast response