Learn More
Hach Company Sension pH combination stainl
Catalog No. 5000207320 Shop All Hach Company Products
Encompass
Sension Plus pH combination stainless steel puncture electrode 5231 for Food Applications pH combination puncture electrode 5231 with one ceramic and one open diaphragm and solid electrolyte Stainless steel body Range 2-14 pH at 0-80C S7 screw cap This electrode is suitable for semisolid food samples e g cheeses meats fish fruits bread doughs etc The encapsulated reference ensures high stability and long lifetime
Catalog No. 50-002-07320 Supplier Hach Company Supplier No. LZW5231.97.0002
Sension Plus pH combination stainless steel puncture electrode 5231 for Food Applications pH combination puncture electrode 5231 with one ceramic and one open diaphragm and solid electrolyte Stainless steel body Range 2-14 pH at 0-80C S7 screw cap This electrode is suitable for semisolid food samples e g cheeses meats fish fruits bread doughs etc The encapsulated reference ensures high stability and long lifetime