Learn More
Hach Company Sension Robust conductivity
Catalog No. 5000207641 Shop All Hach Company Products
Encompass
Sension Plus Robust conductivity cell for field use Robust conductivity cell with Titanium electrodes and Polycarbonat body for Sension Plus portable meters Cell constant k is 1 cm-1 Built-in Pt1000 for automatic temperature compensation 1 m cable MP5 connector Measuring range 5 - 50 micro siemens/cm 0 - 80C Robust conductivity cell for general aqueous application with attractive price position
Catalog No. 50-002-07641 Supplier Hach Company Supplier No. LZW5064.97.0002
Sension Plus Robust conductivity cell for field use Robust conductivity cell with Titanium electrodes and Polycarbonat body for Sension Plus portable meters Cell constant k is 1 cm-1 Built-in Pt1000 for automatic temperature compensation 1 m cable MP5 connector Measuring range 5 - 50 micro siemens/cm 0 - 80C Robust conductivity cell for general aqueous application with attractive price position