Learn More
Hach Company Sension-standard redox solut
Supplier:  Hach Company LZW9400.99
Encompass
Sension Plus-standard redox solution-220 mV-250ml Standard for calibrating and checking accuracy of ORP/Redox electrodes 250mL bottle
Catalog No. 50-002-06962