Learn More
Hach Company Sension-standard redox solut
Supplier:  Hach Company LZW9401.99
Encompass
Sension Plus-standard redox solution-220 mV-1000ml Standard for calibrating and checking accuracy of ORP/Redox electrodes 1L bottle
Catalog No. 50-002-06971