Learn More
ATLANTIC BIOPHARM SOLUTIONS LLC SENSITECH TT ULTRA 16K AMB
Supplier:  ATLANTIC BIOPHARM SOLUTIONS LLC TUA2047071
Encompass_Preferred
Sensitech TT ULTRA 16K AMB 2 DEC SU - MERCK CTS000029SINGLE USE, TempTale Ultra -95°C to +30°C (-139°F to +86°F) rangeLow temp: 1.5 degrees CHigh temp : 8.44 degrees CMeasurement interval: 10 minutes, 1 hour start up delay
Catalog No. NC3805842