Learn More
ATLANTIC BIOPHARM SOLUTIONS LLC SENSITECH TT ULTRA DI AMB 2
Catalog No. NC3805847
Encompass_Preferred
Sensitech TT ULTRA DI AMB 2 DEC SU - MERCK CTS000M20Low temp -90 C, high temp -19.5 C, 10 minute measurement interval, 1.5 hour start up delay
Catalog No. NC3805847 Supplier ATLANTIC BIOPHARM SOLUTIONS LLC Supplier No. TUD2003071
Sensitech TT ULTRA DI AMB 2 DEC SU - MERCK CTS000M20Low temp -90 C, high temp -19.5 C, 10 minute measurement interval, 1.5 hour start up delay